Search
Search

Cmah| Gene Symbol: Cmah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642803.1 | XP_007640993.1 | 14 - 15 | 1881 | 81 | |
| XM_007642804.1 | XP_007640994.1 | 11 - 12 | 1481 | 81 | ||
| XM_007642805.1 | XP_007640995.1 | 9 - 10 | 1182 | 81 | ||
| XM_007642806.1 | XP_007640996.1 | 10 - 11 | 1375 | 81 | ||
| GenBank mRNA | AJ242835.1 | - | 10 - 11 | 1411 | 81 | |
Set Membership: |
|