Search
Search

Rusc1| Gene Symbol: Rusc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501581.2 | XP_003501629.1 | 6 - 7 | 1953 | 70 | |
| XM_007642232.1 | XP_007640422.1 | 5 - 6 | 827 | 70 | ||
| XM_007642233.1 | XP_007640423.1 | 5 - 6 | 590 | 70 | ||
| XM_007642234.1 | XP_007640424.1 | 4 - 5 | 474 | 70 | ||
| XM_007642235.1 | XP_007640425.1 | 5 - 6 | 546 | 70 | ||
| XM_007642236.1 | XP_007640426.1 | 5 - 6 | 501 | 70 | ||
| GenBank mRNA | JI882374.1 | - | 3 - 4 | 365 | 70 | |
| JP058444.1 | - | 3 - 4 | 365 | 70 | ||
Set Membership: |
|