Search
Search

Zfyve20| Gene Symbol: Zfyve20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495175.2 | XP_003495223.1 | 8 - 9 | 1235 | 77 | |
| GenBank mRNA | JI893171.1 | - | 5 - 6 | 548 | 77 | |
| JP050547.1 | - | 5 - 6 | 548 | 77 | ||