Search
Search

LOC100755821| Gene Symbol: LOC100755821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497303.2 | XP_003497351.1 | 5 - 6 | 545 | 97 | |
| XM_007610144.1 | XP_007608334.1 | 5 - 6 | 626 | 97 | ||
| GenBank mRNA | JI877184.1 | - | 5 - 6 | 545 | 97 | |
| JP062646.1 | - | 5 - 6 | 512 | 97 | ||