Search
Search

LOC100772044| Gene Symbol: LOC100772044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498526.2 | XP_003498574.1 | 3 - 4 | 1770 | 106 | |
| GenBank mRNA | JP046949.1 | - | 3 - 4 | 339 | 106 | |
Set Membership: |
|