Search
Search

Atp7a| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505299.2 | XP_003505347.1 | 10 - 11 | 2418 | 86 | |
| XM_007645821.1 | XP_007644011.1 | 10 - 11 | 2447 | 86 | ||
| GenBank mRNA | JI862489.1 | - | 3 - 4 | 479 | 86 | |
| JP055780.1 | - | 3 - 4 | 479 | 86 | ||
Set Membership: |
|