Search
Search

Psmg1| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508729.2 | XP_003508777.1 | 4 - 5 | 501 | 80 | |
| GenBank mRNA | JI881292.1 | - | 1 - 2 | 63 | 80 | |
| JP037183.1 | - | 1 - 2 | 63 | 80 | ||
Set Membership: |
|