Search
Search

Stxbp5| Gene Symbol: Stxbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498949.2 | XP_003498997.1 | 17 - 18 | 2005 | 76 | |
| XM_003498950.2 | XP_003498998.1 | 17 - 18 | 2005 | 76 | ||
| XM_003498951.2 | XP_003498999.1 | 17 - 18 | 2005 | 76 | ||
| GenBank mRNA | JI863821.1 | - | 15 - 16 | 1681 | 76 | |
| JP048303.1 | - | 15 - 16 | 1681 | 76 | ||
Set Membership: |
|