Search
Search

Plekhg4| Gene Symbol: Plekhg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649170.1 | XP_007647360.1 | 8 - 9 | 1156 | 62 | |
| XM_007649171.1 | XP_007647361.1 | 8 - 9 | 1415 | 62 | ||
| XM_007649172.1 | XP_007647362.1 | 7 - 8 | 1220 | 62 | ||
| XM_007649173.1 | XP_007647363.1 | 8 - 9 | 981 | 62 | ||
Set Membership: |
|