Search
Search

Sox6| Gene Symbol: Sox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504806.2 | XP_003504854.1 | 15 - 16 | 2331 | 57 | |
| XM_007645305.1 | XP_007643495.1 | 15 - 16 | 2261 | 57 | ||
| XM_007645306.1 | XP_007643496.1 | 15 - 16 | 2325 | 57 | ||
| XM_007645307.1 | XP_007643497.1 | 13 - 14 | 1784 | 57 | ||
| XM_007645308.1 | XP_007643498.1 | 12 - 13 | 1754 | 57 | ||
| XM_007645310.1 | XP_007643500.1 | 14 - 15 | 2208 | 57 | ||
Set Membership: |
|