Search
Search

Pnpla2| Gene Symbol: Pnpla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509767.2 | XP_003509815.1 | 8 - 9 | 1298 | 65 | |
| GenBank mRNA | JI869961.1 | - | 8 - 9 | 1284 | 65 | |
| JP060132.1 | - | 8 - 9 | 1283 | 65 | ||
Set Membership: |
|