Search
Search

Frem1| Gene Symbol: Frem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642351.1 | XP_007640541.1 | 7 - 8 | 1418 | 60 | |
| XM_007642353.1 | XP_007640543.1 | 5 - 6 | 1124 | 60 | ||
| XM_007642354.1 | XP_007640544.1 | 3 - 4 | 705 | 60 | ||
Set Membership: |
|