Search
Search

Phactr1| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497940.2 | XP_003497988.1 | 9 - 10 | 1552 | 69 | |
| XM_007619938.1 | XP_007618128.1 | 1 - 2 | 133 | 69 | ||
Set Membership: |
|