Search
Search

Sgol2| Gene Symbol: Sgol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642652.1 | XP_007640842.1 | 4 - 5 | 645 | 94 | |
| XM_007642653.1 | XP_007640843.1 | 4 - 5 | 541 | 94 | ||
| GenBank mRNA | JI884768.1 | - | 3 - 4 | 376 | 94 | |
| JP046864.1 | - | 3 - 4 | 375 | 94 | ||
Set Membership: |
|