Search
Search

Atic| Gene Symbol: Atic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649845.1 | XP_007648035.1 | 5 - 6 | 481 | 57 | |
| GenBank mRNA | JI875543.1 | - | 5 - 6 | 496 | 57 | |
| JP061757.1 | - | 5 - 6 | 496 | 57 | ||
Set Membership: |
|