Search
Search

Slc24a1| Gene Symbol: Slc24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507140.2 | XP_003507188.1 | 7 - 8 | 3029 | 65 | |
| XM_003507141.2 | XP_003507189.1 | 7 - 8 | 2657 | 65 | ||
| XM_007647704.1 | XP_007645894.1 | 8 - 9 | 2879 | 65 | ||
Set Membership: |
|