Search
Search

Sel1l3| Gene Symbol: Sel1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512097.2 | XP_003512145.2 | 14 - 15 | 2141 | 61 | |
| GenBank mRNA | JI874091.1 | - | 2 - 3 | 179 | 61 | |
| JP062754.1 | - | 2 - 3 | 179 | 61 | ||
Set Membership: |
|