Search
Search

Fgf13| Gene Symbol: Fgf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641948.1 | XP_007640138.1 | 5 - 6 | 643 | 70 | |
| XM_007641949.1 | XP_007640139.1 | 5 - 6 | 671 | 70 | ||
| GenBank mRNA | JI896328.1 | - | 1 - 2 | 117 | 70 | |
Set Membership: |
|