Search
Search

Rabep1| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608875.1 | XP_007607065.1 | 11 - 12 | 1853 | 60 | |
| XM_007608881.1 | XP_007607071.1 | 11 - 12 | 1832 | 60 | ||
| XM_007608887.1 | XP_007607077.1 | 10 - 11 | 1748 | 60 | ||
| GenBank mRNA | JI875624.1 | - | 3 - 4 | 247 | 60 | |
| JP048624.1 | - | 3 - 4 | 247 | 60 | ||
Set Membership: |
|