Search
Search

Znf143| Gene Symbol: Znf143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511181.2 | XP_003511229.1 | 9 - 10 | 936 | 79 | |
| XM_007651724.1 | XP_007649914.1 | 9 - 10 | 921 | 79 | ||
| XM_007651725.1 | XP_007649915.1 | 9 - 10 | 918 | 79 | ||
| XM_007651726.1 | XP_007649916.1 | 9 - 10 | 931 | 79 | ||
| GenBank mRNA | JI869507.1 | - | 4 - 5 | 419 | 79 | |
| JP049627.1 | - | 4 - 5 | 419 | 79 | ||
Set Membership: |
|