Search
Search

Mterfd1| Gene Symbol: Mterfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511573.2 | XP_003511621.1 | 7 - 8 | 1078 | 95 | |
| GenBank mRNA | JI896737.1 | - | 6 - 7 | 1102 | 95 | |
| JP060486.1 | - | 6 - 7 | 1102 | 95 | ||
Set Membership: |
|