Search
Search

Mdfic| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650914.1 | XP_007649104.1 | 3 - 4 | 449 | 111 | |
| XM_007650916.1 | XP_007649106.1 | 3 - 4 | 675 | 111 | ||
| GenBank mRNA | JI873000.1 | - | 1 - 2 | 264 | 111 | |
Set Membership: |
|