Search
Search

Poc1b| Gene Symbol: Poc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644865.1 | XP_007643055.1 | 9 - 10 | 1220 | 69 | |
| GenBank mRNA | JI890841.1 | - | 9 - 10 | 1341 | 69 | |
| JP054296.1 | - | 9 - 10 | 1341 | 69 | ||
Set Membership: |
|