Search
Search

Lman1l| Gene Symbol: Lman1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514372.1 | XP_003514420.1 | 7 - 8 | 776 | 63 | |
| XM_007654521.1 | XP_007652711.1 | 7 - 8 | 776 | 63 | ||
Set Membership: |
|