Search
Search

Tyk2| Gene Symbol: Tyk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499738.2 | XP_003499786.1 | 7 - 8 | 1285 | 56 | |
| XM_007640494.1 | XP_007638684.1 | 8 - 9 | 1567 | 56 | ||
| XM_007640495.1 | XP_007638685.1 | 8 - 9 | 1437 | 56 | ||
| XM_007640496.1 | XP_007638686.1 | 8 - 9 | 1440 | 56 | ||
| XM_007640497.1 | XP_007638687.1 | 7 - 8 | 1333 | 56 | ||
| XM_007640498.1 | XP_007638688.1 | 7 - 8 | 1330 | 56 | ||
| XM_007640499.1 | XP_007638689.1 | 4 - 5 | 780 | 56 | ||
Set Membership: |
|