Search
Search

Edrf1| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513870.2 | XP_003513918.1 | 21 - 22 | 3244 | 58 | |
| XM_007654101.1 | XP_007652291.1 | 21 - 22 | 3244 | 58 | ||
Set Membership: |
|