Search
Search

Xrcc6| Gene Symbol: Xrcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514729.2 | XP_003514777.1 | 11 - 12 | 1609 | 67 | |
| XM_007654785.1 | XP_007652975.1 | 9 - 10 | 1439 | 67 | ||
| GenBank mRNA | JI896268.1 | - | 11 - 12 | 1617 | 67 | |
| JP040686.1 | - | 11 - 12 | 1617 | 67 | ||
Set Membership: |
|