Search
Search

Dbndd2| Gene Symbol: Dbndd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645109.1 | XP_007643299.1 | 3 - 4 | 832 | 64 | |
| XM_007645110.1 | XP_007643300.1 | 3 - 4 | 810 | 64 | ||
| XM_007645111.1 | XP_007643301.1 | 3 - 4 | 784 | 64 | ||
| XM_007645112.1 | XP_007643302.1 | 2 - 3 | 286 | 64 | ||
| XM_007645113.1 | XP_007643303.1 | 3 - 4 | 430 | 64 | ||
| XM_007645114.1 | XP_007643304.1 | 3 - 4 | 480 | 64 | ||
| GenBank mRNA | JP063969.1 | - | 2 - 3 | 267 | 64 | |
Set Membership: |
|