Search
Search

Cc2d1a| Gene Symbol: Cc2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501932.2 | XP_003501980.1 | 22 - 23 | 2501 | 54 | |
| GenBank mRNA | JI863798.1 | - | 5 - 6 | 375 | 54 | |
| JP049983.1 | - | 5 - 6 | 375 | 54 | ||
Set Membership: |
|