Search
Search

Polr3g| Gene Symbol: Polr3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504392.2 | XP_003504440.1 | 5 - 6 | 404 | 65 | |
| XM_007644916.1 | XP_007643106.1 | 4 - 5 | 389 | 65 | ||
| GenBank mRNA | JI896141.1 | - | 4 - 5 | 384 | 65 | |
| JP055460.1 | - | 4 - 5 | 384 | 65 | ||
Set Membership: |
|