Search
Search

Ppp3cc| Gene Symbol: Ppp3cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647554.1 | XP_007645744.1 | 11 - 12 | 1287 | 63 | |
| XM_007647558.1 | XP_007645748.1 | 11 - 12 | 1286 | 63 | ||
| XM_007647562.1 | XP_007645752.1 | 11 - 12 | 1287 | 63 | ||
| GenBank mRNA | JI881035.1 | - | 10 - 11 | 1011 | 63 | |
Set Membership: |
|