Search
Search

Trmt2a| Gene Symbol: Trmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502453.2 | XP_003502501.1 | 2 - 3 | 895 | 57 | |
| XM_003502454.2 | XP_003502502.1 | 2 - 3 | 847 | 57 | ||
| GenBank mRNA | JI878075.1 | - | 2 - 3 | 877 | 57 | |
| JP048452.1 | - | 2 - 3 | 877 | 57 | ||
Set Membership: |
|