Search
Search

Hectd2| Gene Symbol: Hectd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007636157.1 | XP_007634347.1 | 3 - 4 | 302 | 67 | |
| GenBank mRNA | JI867453.1 | - | 3 - 4 | 462 | 67 | |
| JP044796.1 | - | 3 - 4 | 462 | 67 | ||
Set Membership: |
|