Search
Search

Itgb1| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649040.1 | XP_007647230.1 | 6 - 7 | 957 | 81 | |
| GenBank mRNA | JI884008.1 | - | 6 - 7 | 1011 | 81 | |
| JP066108.1 | - | 6 - 7 | 1011 | 81 | ||
Set Membership: |
|