Search
Search

Col16a1| Gene Symbol: Col16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007626445.1 | XP_007624635.1 | 46 - 47 | 3271 | 51 | |
| XM_007626453.1 | XP_007624643.1 | 46 - 47 | 3268 | 51 | ||
| XM_007626457.1 | XP_007624647.1 | 45 - 46 | 3220 | 51 | ||
| XM_007626465.1 | XP_007624655.1 | 45 - 46 | 3220 | 51 | ||
| XM_007626467.1 | XP_007624657.1 | 44 - 45 | 3113 | 51 | ||
Set Membership: |
|