Search
Search

Abcd4| Gene Symbol: Abcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514268.2 | XP_003514316.1 | 18 - 19 | 1808 | 80 | |
| GenBank mRNA | JI890642.1 | - | 6 - 7 | 391 | 80 | |
| JP058224.1 | - | 6 - 7 | 391 | 80 | ||
Set Membership: |
|