Search
Search

Mmp21| Gene Symbol: Mmp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513871.2 | XP_003513919.1 | 6 - 7 | 1905 | 111 | |
| XM_007654102.1 | XP_007652292.1 | 6 - 7 | 1923 | 111 | ||
Set Membership: |
|