Search
Search

Mynn| Gene Symbol: Mynn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503502.2 | XP_003503550.1 | 4 - 5 | 2188 | 103 | |
| XM_007644110.1 | XP_007642300.1 | 4 - 5 | 2187 | 103 | ||
| XM_007644111.1 | XP_007642301.1 | 3 - 4 | 1085 | 103 | ||
| XM_007644112.1 | XP_007642302.1 | 4 - 5 | 1052 | 103 | ||
| GenBank mRNA | JI869512.1 | - | 2 - 3 | 952 | 103 | |
| JP041229.1 | - | 2 - 3 | 952 | 103 | ||
Set Membership: |
|