Search
Search

Tspan12| Gene Symbol: Tspan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507394.2 | XP_003507442.1 | 2 - 3 | 158 | 65 | |
| XM_007647994.1 | XP_007646184.1 | 2 - 3 | 384 | 65 | ||
| XM_007647995.1 | XP_007646185.1 | 2 - 3 | 134 | 65 | ||
Set Membership: |
|