Search
Search

Ces3| Gene Symbol: Ces3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508532.1 | XP_003508580.1 | 10 - 11 | 1288 | 80 | |
| XM_003508533.1 | XP_003508581.1 | 9 - 10 | 1147 | 80 | ||
Set Membership: |
|