Search
Search

Col8a1| Gene Symbol: Col8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504870.2 | XP_003504918.1 | 3 - 4 | 1024 | 83 | |
| XM_007645368.1 | XP_007643558.1 | 4 - 5 | 1178 | 83 | ||
Set Membership: |
|