Search
Search

LOC100770001| Gene Symbol: LOC100770001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509818.2 | XP_003509866.1 | 6 - 7 | 1232 | 90 | |
| XM_007650463.1 | XP_007648653.1 | 5 - 6 | 1786 | 90 | ||
| XM_007650464.1 | XP_007648654.1 | 6 - 7 | 956 | 90 | ||
| XM_007650465.1 | XP_007648655.1 | 6 - 7 | 920 | 90 | ||
| GenBank mRNA | JI865689.1 | - | 5 - 6 | 795 | 90 | |
| JP060556.1 | - | 6 - 7 | 1181 | 90 | ||