Search
Search

Kmt2a| Gene Symbol: Kmt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651151.1 | XP_007649341.1 | 30 - 31 | 10684 | 96 | |
| GenBank mRNA | JI895186.1 | - | 3 - 4 | 2221 | 96 | |
| JP047379.1 | - | 3 - 4 | 2185 | 96 | ||
Set Membership: |
|