Search
Search

Terf2ip| Gene Symbol: Terf2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510157.2 | XP_003510205.1 | 1 - 2 | 765 | 94 | |
| XM_007650796.1 | XP_007648986.1 | 1 - 2 | 766 | 94 | ||
| GenBank mRNA | JI877703.1 | - | 1 - 2 | 170 | 94 | |
| JP052141.1 | - | 1 - 2 | 170 | 94 | ||
Set Membership: |
|