Search
Search

Magt1| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505297.2 | XP_003505345.1 | 9 - 10 | 1037 | 91 | |
| GenBank mRNA | JI885607.1 | - | 9 - 10 | 1037 | 91 | |
| JP055298.1 | - | 9 - 10 | 1037 | 91 | ||
Set Membership: |
|