Search
Search

Swap70| Gene Symbol: Swap70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511182.2 | XP_003511230.1 | 11 - 12 | 1749 | 71 | |
| GenBank mRNA | JI883881.1 | - | 2 - 3 | 297 | 71 | |
| JP062251.1 | - | 2 - 3 | 297 | 71 | ||
Set Membership: |
|