Search
Search

Lztfl1| Gene Symbol: Lztfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648313.1 | XP_007646503.1 | 6 - 7 | 714 | 80 | |
| XM_007648314.1 | XP_007646504.1 | 6 - 7 | 580 | 80 | ||
| XM_007648315.1 | XP_007646505.1 | 6 - 7 | 544 | 80 | ||
| XM_007648316.1 | XP_007646506.1 | 6 - 7 | 676 | 80 | ||
| GenBank mRNA | JI893335.1 | - | 5 - 6 | 548 | 80 | |
Set Membership: |
|