Search
Search

Dcaf5| Gene Symbol: Dcaf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007625137.1 | XP_007623327.1 | 7 - 8 | 891 | 90 | |
| XM_007625146.1 | XP_007623336.1 | 4 - 5 | 504 | 90 | ||
| GenBank mRNA | JI867570.1 | - | 7 - 8 | 953 | 90 | |
| JP051395.1 | - | 7 - 8 | 953 | 90 | ||
Set Membership: |
|