Search
Search

Bmpr1a| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495327.2 | XP_003495375.1 | 5 - 6 | 648 | 64 | |
| XM_007638484.1 | XP_007636674.1 | 5 - 6 | 650 | 64 | ||
| GenBank mRNA | JI889293.1 | - | 1 - 2 | 77 | 64 | |
| JP060440.1 | - | 1 - 2 | 77 | 64 | ||
Set Membership: |
|