Search
Search

LOC100751499| Gene Symbol: LOC100751499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512386.2 | XP_003512434.1 | 6 - 7 | 895 | 65 | |
| GenBank mRNA | JI862118.1 | - | 4 - 5 | 564 | 65 | |
| JP048190.1 | - | 4 - 5 | 564 | 65 | ||
Set Membership: |
|